Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604475 | ||||||||||||||||||||
Literature Links: |
EML6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EML6 - echinoderm microtubule associated protein like 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039753.2 | 894 | Intron | NP_001034842.2 | |||
XM_017004098.1 | 894 | Intron | XP_016859587.1 | |||
XM_017004099.1 | 894 | Intron | XP_016859588.1 | |||
XM_017004100.1 | 894 | Intron | XP_016859589.1 | |||
XM_017004101.1 | 894 | Intron | XP_016859590.1 | |||
XM_017004102.1 | 894 | Intron | XP_016859591.1 |
RTN4 - reticulon 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321859.1 | 894 | UTR 3 | NP_001308788.1 | |||
NM_001321860.1 | 894 | UTR 3 | NP_001308789.1 | |||
NM_001321861.1 | 894 | UTR 3 | NP_001308790.1 | |||
NM_001321862.1 | 894 | UTR 3 | NP_001308791.1 | |||
NM_001321863.1 | 894 | UTR 3 | NP_001308792.1 | |||
NM_001321904.1 | 894 | UTR 3 | NP_001308833.1 | |||
NM_007008.2 | 894 | UTR 3 | NP_008939.1 | |||
NM_020532.4 | 894 | UTR 3 | NP_065393.1 | |||
NM_153828.2 | 894 | UTR 3 | NP_722550.1 | |||
NM_207520.1 | 894 | UTR 3 | NP_997403.1 | |||
NM_207521.1 | 894 | UTR 3 | NP_997404.1 | |||
XM_005264434.3 | 894 | UTR 3 | XP_005264491.1 | |||
XM_017004518.1 | 894 | UTR 3 | XP_016860007.1 | |||
XM_017004519.1 | 894 | UTR 3 | XP_016860008.1 |