Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTTTTCTCCAAGAATCCAATATT[C/T]CCAGTATATTTTCATATACTTTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603310 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC645513 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC645513 - septin 7 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDE5A - phosphodiesterase 5A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083.3 | 5582 | UTR 3 | NP_001074.2 | |||
NM_033430.2 | 5582 | UTR 3 | NP_236914.2 | |||
NM_033437.3 | 5582 | UTR 3 | NP_246273.2 | |||
XM_017008791.1 | 5582 | Intron | XP_016864280.1 |
Set Membership: |
HapMap Validated |