Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTAAATTGCTCATATAGTTTAGTC[C/T]ACAGAGACAAGATTTTTCTCTTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608019 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TNIP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TNIP3 - TNFAIP3 interacting protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128843.2 | Intron | NP_001122315.2 | ||||
NM_001244764.1 | Intron | NP_001231693.1 | ||||
NM_024873.5 | Intron | NP_079149.3 | ||||
XM_011532256.2 | Intron | XP_011530558.1 | ||||
XM_011532257.2 | Intron | XP_011530559.1 | ||||
XM_017008619.1 | Intron | XP_016864108.1 | ||||
XM_017008620.1 | Intron | XP_016864109.1 | ||||
XM_017008621.1 | Intron | XP_016864110.1 | ||||
XM_017008622.1 | Intron | XP_016864111.1 | ||||
XM_017008623.1 | Intron | XP_016864112.1 | ||||
XM_017008624.1 | Intron | XP_016864113.1 | ||||
XM_017008625.1 | Intron | XP_016864114.1 |