Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCATAGTGAAATAAGAATATTCA[A/G]TATAAATTTTGAGTTTAACTTTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
YTHDF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
YTHDF3 - YTH N6-methyladenosine RNA binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277813.1 | Intron | NP_001264742.1 | ||||
NM_001277814.1 | Intron | NP_001264743.1 | ||||
NM_001277815.1 | Intron | NP_001264744.1 | ||||
NM_001277816.1 | Intron | NP_001264745.1 | ||||
NM_001277817.1 | Intron | NP_001264746.1 | ||||
NM_001277818.1 | Intron | NP_001264747.1 | ||||
NM_152758.5 | Intron | NP_689971.4 | ||||
XM_011517509.2 | Intron | XP_011515811.1 | ||||
XM_011517510.2 | Intron | XP_011515812.1 |