Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTCAAATGTAGTGTTTTCTTTTT[C/T]TAAGACATTGCAGATTGAATTCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
YTHDF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
YTHDF3 - YTH N6-methyladenosine RNA binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277813.1 | Intron | NP_001264742.1 | ||||
NM_001277814.1 | Intron | NP_001264743.1 | ||||
NM_001277815.1 | Intron | NP_001264744.1 | ||||
NM_001277816.1 | Intron | NP_001264745.1 | ||||
NM_001277817.1 | Intron | NP_001264746.1 | ||||
NM_001277818.1 | Intron | NP_001264747.1 | ||||
NM_152758.5 | Intron | NP_689971.4 | ||||
XM_011517509.2 | Intron | XP_011515811.1 | ||||
XM_011517510.2 | Intron | XP_011515812.1 |
YTHDF3-AS1 - YTHDF3 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |