Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
NAA40 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NAA40 - N(alpha)-acetyltransferase 40, NatD catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300800.1 | 33 | Intron | NP_001287729.1 | |||
NM_024771.3 | 33 | UTR 5 | NP_079047.2 | |||
XM_006718689.3 | 33 | UTR 5 | XP_006718752.1 | |||
XM_011545254.2 | 33 | Silent Mutation | CCC,CCG | P,P 6 | XP_011543556.1 | |
XM_011545255.2 | 33 | Silent Mutation | CCC,CCG | P,P 6 | XP_011543557.1 |