Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164014 MIM: 602180 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR4489 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MIR4489 - microRNA 4489 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RELA - RELA proto-oncogene, NF-kB subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145138.1 | 2296 | UTR 3 | NP_001138610.1 | |||
NM_001243984.1 | 2296 | UTR 3 | NP_001230913.1 | |||
NM_001243985.1 | 2296 | UTR 3 | NP_001230914.1 | |||
NM_021975.3 | 2296 | UTR 3 | NP_068810.3 | |||
XM_011545206.1 | 2296 | UTR 3 | XP_011543508.1 | |||
XM_011545207.1 | 2296 | UTR 3 | XP_011543509.1 |
SIPA1 - signal-induced proliferation-associated 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |