Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613368 MIM: 609849 MIM: 601577 MIM: 608939 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CARNS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CARNS1 - carnosine synthase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CORO1B - coronin 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTPRCAP - protein tyrosine phosphatase, receptor type C associated protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPS6KB2 - ribosomal protein S6 kinase B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003952.2 | Intron | NP_003943.2 | ||||
XM_005274164.1 | Intron | XP_005274221.1 | ||||
XM_005274165.4 | Intron | XP_005274222.1 | ||||
XM_006718655.3 | Intron | XP_006718718.1 | ||||
XM_006718656.3 | Intron | XP_006718719.1 | ||||
XM_006718657.1 | Intron | XP_006718720.1 | ||||
XM_017018108.1 | Intron | XP_016873597.1 |
Set Membership: |
HapMap |