Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600956 MIM: 189906 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMHR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AMHR2 - anti-Mullerian hormone receptor type 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164690.1 | Intron | NP_001158162.1 | ||||
NM_001164691.1 | Intron | NP_001158163.1 | ||||
NM_020547.2 | Intron | NP_065434.1 | ||||
XM_011538173.1 | Intron | XP_011536475.1 | ||||
XM_011538174.1 | Intron | XP_011536476.1 | ||||
XM_011538176.1 | Intron | XP_011536478.1 | ||||
XM_011538178.1 | Intron | XP_011536480.1 | ||||
XM_011538179.1 | Intron | XP_011536481.1 | ||||
XM_011538180.1 | Intron | XP_011536482.1 | ||||
XM_011538181.1 | Intron | XP_011536483.1 | ||||
XM_011538183.2 | Intron | XP_011536485.1 | ||||
XM_011538184.2 | Intron | XP_011536486.1 | ||||
XM_011538185.1 | Intron | XP_011536487.1 | ||||
XM_011538186.2 | Intron | XP_011536488.1 | ||||
XM_011538187.2 | Intron | XP_011536489.1 | ||||
XM_017019179.1 | Intron | XP_016874668.1 |
SP1 - Sp1 transcription factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |