Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600447 MIM: 601210 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP3K12 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
MAP3K12 - mitogen-activated protein kinase kinase kinase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193511.1 | Intron | NP_001180440.1 | ||||
NM_006301.3 | Intron | NP_006292.3 | ||||
XM_005269138.3 | Intron | XP_005269195.1 | ||||
XM_005269140.2 | Intron | XP_005269197.1 | ||||
XM_006719588.3 | Intron | XP_006719651.1 | ||||
XM_011538725.2 | Intron | XP_011537027.1 | ||||
XM_017019956.1 | Intron | XP_016875445.1 |
PCBP2 - poly(rC) binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098620.2 | Intron | NP_001092090.1 | ||||
NM_001128911.1 | Intron | NP_001122383.1 | ||||
NM_001128912.1 | Intron | NP_001122384.1 | ||||
NM_001128913.1 | Intron | NP_001122385.1 | ||||
NM_001128914.1 | Intron | NP_001122386.1 | ||||
NM_005016.5 | Intron | NP_005007.2 | ||||
NM_031989.4 | Intron | NP_114366.1 |
Set Membership: |
HapMap Validated |