Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606371 MIM: 600447 MIM: 604643 MIM: 605053 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATF7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ATF7 - activating transcription factor 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAP3K12 - mitogen-activated protein kinase kinase kinase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193511.1 | 386 | Intron | NP_001180440.1 | |||
NM_006301.3 | 386 | Intron | NP_006292.3 | |||
XM_005269138.3 | 386 | Intron | XP_005269195.1 | |||
XM_005269140.2 | 386 | Intron | XP_005269197.1 | |||
XM_006719588.3 | 386 | Intron | XP_006719651.1 | |||
XM_011538725.2 | 386 | UTR 5 | XP_011537027.1 | |||
XM_017019956.1 | 386 | Intron | XP_016875445.1 |
NPFF - neuropeptide FF-amide peptide precursor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TARBP2 - TARBP2, RISC loading complex RNA binding subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004178.4 | 386 | Intron | NP_004169.3 | |||
NM_134323.1 | 386 | Intron | NP_599150.1 | |||
NM_134324.2 | 386 | Intron | NP_599151.2 | |||
XM_005269114.1 | 386 | Intron | XP_005269171.1 | |||
XM_005269115.2 | 386 | Intron | XP_005269172.1 | |||
XM_005269117.1 | 386 | Intron | XP_005269174.1 | |||
XM_005269120.4 | 386 | Intron | XP_005269177.1 | |||
XM_005269122.3 | 386 | Intron | XP_005269179.1 | |||
XM_006719581.1 | 386 | Intron | XP_006719644.1 | |||
XM_011538712.2 | 386 | Intron | XP_011537014.1 | |||
XM_017019910.1 | 386 | Intron | XP_016875399.1 | |||
XM_017019911.1 | 386 | Intron | XP_016875400.1 |
Set Membership: |
HapMap |