Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
69 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164690 MIM: 607555 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABL2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ABL2 - ABL proto-oncogene 2, non-receptor tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136000.2 | 8112 | UTR 3 | NP_001129472.1 | |||
NM_001136001.1 | 8112 | Intron | NP_001129473.1 | |||
NM_001168236.1 | 8112 | UTR 3 | NP_001161708.1 | |||
NM_001168237.1 | 8112 | UTR 3 | NP_001161709.1 | |||
NM_001168238.1 | 8112 | UTR 3 | NP_001161710.1 | |||
NM_001168239.1 | 8112 | UTR 3 | NP_001161711.1 | |||
NM_005158.4 | 8112 | UTR 3 | NP_005149.4 | |||
NM_007314.3 | 8112 | UTR 3 | NP_009298.1 | |||
XM_005245088.2 | 8112 | Intron | XP_005245145.1 | |||
XM_017001035.1 | 8112 | Intron | XP_016856524.1 |
TOR3A - torsin family 3 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |