Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
53 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600497 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf168 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1orf168 - chromosome 1 open reading frame 168 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004303.4 | Intron | NP_001004303.3 | ||||
XM_005270584.3 | Intron | XP_005270641.1 | ||||
XM_011540898.1 | Intron | XP_011539200.1 | ||||
XM_011540899.1 | Intron | XP_011539201.1 | ||||
XM_011540900.2 | Intron | XP_011539202.1 | ||||
XM_011540903.2 | Intron | XP_011539205.1 | ||||
XM_011540904.2 | Intron | XP_011539206.1 | ||||
XM_011540905.2 | Intron | XP_011539207.1 | ||||
XM_011540906.2 | Intron | XP_011539208.1 | ||||
XM_017000544.1 | Intron | XP_016856033.1 | ||||
XM_017000545.1 | Intron | XP_016856034.1 | ||||
XM_017000546.1 | Intron | XP_016856035.1 | ||||
XM_017000547.1 | Intron | XP_016856036.1 | ||||
XM_017000548.1 | Intron | XP_016856037.1 |
PRKAA2 - protein kinase AMP-activated catalytic subunit alpha 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
JSNP |