Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCCTTGCAGAAAAAAATAACCTC[A/G]CAGCTTTGAAAAAGATCTGGGATCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612786 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCNY PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CCNY - cyclin Y | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282852.1 | Intron | NP_001269781.1 | ||||
NM_001282853.1 | Intron | NP_001269782.1 | ||||
NM_001282854.1 | Intron | NP_001269783.1 | ||||
NM_145012.5 | Intron | NP_659449.3 | ||||
NM_181698.3 | Intron | NP_859049.2 | ||||
XM_006717392.3 | Intron | XP_006717455.1 | ||||
XM_011519357.2 | Intron | XP_011517659.1 | ||||
XM_011519358.1 | Intron | XP_011517660.1 | ||||
XM_011519360.2 | Intron | XP_011517662.1 | ||||
XM_011519361.1 | Intron | XP_011517663.1 | ||||
XM_011519362.1 | Intron | XP_011517664.1 | ||||
XM_011519363.1 | Intron | XP_011517665.1 | ||||
XM_011519364.1 | Intron | XP_011517666.1 | ||||
XM_011519365.2 | Intron | XP_011517667.1 | ||||
XM_011519366.1 | Intron | XP_011517668.1 | ||||
XM_011519367.1 | Intron | XP_011517669.1 | ||||
XM_011519368.2 | Intron | XP_011517670.1 | ||||
XM_011519369.2 | Intron | XP_011517671.1 | ||||
XM_017015839.1 | Intron | XP_016871328.1 | ||||
XM_017015840.1 | Intron | XP_016871329.1 | ||||
XM_017015841.1 | Intron | XP_016871330.1 |