Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615317 MIM: 602091 MIM: 601015 | ||||||||||||||||||||
Literature Links: |
ISCA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ISCA2 - iron-sulfur cluster assembly 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LTBP2 - latent transforming growth factor beta binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000428.2 | 8028 | UTR 3 | NP_000419.1 | |||
XM_011536765.1 | 8028 | UTR 3 | XP_011535067.1 |
NPC2 - NPC intracellular cholesterol transporter 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |