Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 137207 | ||||||||||||||||||||
Literature Links: |
LRRC32 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LRRC32 - leucine rich repeat containing 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128922.1 | 3335 | UTR 3 | NP_001122394.1 | |||
NM_005512.2 | 3335 | UTR 3 | NP_005503.1 | |||
XM_005273902.3 | 3335 | Intron | XP_005273959.1 | |||
XM_005273903.4 | 3335 | UTR 3 | XP_005273960.2 | |||
XM_011544935.2 | 3335 | UTR 3 | XP_011543237.1 | |||
XM_017017527.1 | 3335 | Intron | XP_016873016.1 |