Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604923 | ||||||||||||||||||||
Literature Links: |
MAP4K5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAP4K5 - mitogen-activated protein kinase kinase kinase kinase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006575.4 | 2825 | Missense Mutation | AAA,AAT | K,N 835 | NP_006566.2 | |
NM_198794.2 | 2825 | Missense Mutation | AAA,AAT | K,N 835 | NP_942089.1 | |
XM_006720013.3 | 2825 | Missense Mutation | AAA,AAT | K,N 835 | XP_006720076.1 | |
XM_011536377.2 | 2825 | Missense Mutation | AAA,AAT | K,N 834 | XP_011534679.1 | |
XM_011536378.2 | 2825 | Missense Mutation | AAA,AAT | K,N 768 | XP_011534680.1 | |
XM_011536379.2 | 2825 | Missense Mutation | AAA,AAT | K,N 699 | XP_011534681.1 | |
XM_011536380.2 | 2825 | Missense Mutation | AAA,AAT | K,N 546 | XP_011534682.1 |