Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607512 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAMTS18 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ADAMTS18 - ADAM metallopeptidase with thrombospondin type 1 motif 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001326358.1 | 5199 | UTR 3 | NP_001313287.1 | |||
NM_199355.3 | 5199 | UTR 3 | NP_955387.1 | |||
XM_011522923.1 | 5199 | UTR 3 | XP_011521225.1 | |||
XM_011522924.1 | 5199 | UTR 3 | XP_011521226.1 | |||
XM_017022988.1 | 5199 | UTR 3 | XP_016878477.1 | |||
XM_017022989.1 | 5199 | UTR 3 | XP_016878478.1 |
Set Membership: |
HapMap |