Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605015 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF214 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF214 - zinc finger protein 214 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013249.2 | 2663 | UTR 3 | NP_037381.2 | |||
XM_005253128.3 | 2663 | UTR 3 | XP_005253185.1 | |||
XM_005253129.3 | 2663 | UTR 3 | XP_005253186.1 | |||
XM_006718308.3 | 2663 | UTR 3 | XP_006718371.1 | |||
XM_006718309.3 | 2663 | UTR 3 | XP_006718372.1 | |||
XM_011520355.2 | 2663 | UTR 3 | XP_011518657.1 | |||
XM_011520357.2 | 2663 | UTR 3 | XP_011518659.1 | |||
XM_017018269.1 | 2663 | UTR 3 | XP_016873758.1 |