Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 159430 MIM: 604760 | ||||||||||||||||||||
Literature Links: |
MBP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MBP - myelin basic protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025081.1 | 1846 | UTR 3 | NP_001020252.1 | |||
NM_001025090.1 | 1846 | UTR 3 | NP_001020261.1 | |||
NM_001025092.1 | 1846 | UTR 3 | NP_001020263.1 | |||
NM_001025100.1 | 1846 | Intron | NP_001020271.1 | |||
NM_001025101.1 | 1846 | UTR 3 | NP_001020272.1 | |||
NM_002385.2 | 1846 | UTR 3 | NP_002376.1 | |||
XM_011526009.2 | 1846 | Intron | XP_011524311.1 | |||
XM_017025778.1 | 1846 | Intron | XP_016881267.1 | |||
XM_017025779.1 | 1846 | Intron | XP_016881268.1 | |||
XM_017025780.1 | 1846 | Intron | XP_016881269.1 |
ZNF236 - zinc finger protein 236 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |