Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604996 MIM: 608303 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FXYD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FXYD3 - FXYD domain containing ion transport regulator 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136007.1 | Intron | NP_001129479.1 | ||||
NM_001136008.1 | Intron | NP_001129480.1 | ||||
NM_001136009.1 | Intron | NP_001129481.1 | ||||
NM_001136010.1 | Intron | NP_001129482.1 | ||||
NM_001136011.1 | Intron | NP_001129483.1 | ||||
NM_001136012.1 | Intron | NP_001129484.1 | ||||
NM_005971.3 | Intron | NP_005962.1 | ||||
NM_021910.2 | Intron | NP_068710.1 | ||||
XM_005258996.4 | Intron | XP_005259053.1 | ||||
XM_011527032.2 | Intron | XP_011525334.1 | ||||
XM_017026877.1 | Intron | XP_016882366.1 | ||||
XM_017026878.1 | Intron | XP_016882367.1 |
LGI4 - leucine rich repeat LGI family member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6887 - microRNA 6887 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |