Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602242 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP2S1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
AP2S1 - adaptor related protein complex 2 sigma 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301076.1 | 846 | UTR 3 | NP_001288005.1 | |||
NM_001301078.1 | 846 | UTR 3 | NP_001288007.1 | |||
NM_001301081.1 | 846 | UTR 3 | NP_001288010.1 | |||
NM_004069.4 | 846 | UTR 3 | NP_004060.2 | |||
NM_021575.3 | 846 | UTR 3 | NP_067586.1 | |||
XM_011526423.2 | 846 | UTR 3 | XP_011524725.1 | |||
XM_011526424.2 | 846 | UTR 3 | XP_011524726.1 |
SNAR-E - small ILF3/NF90-associated RNA E | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |