Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602003 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRMP PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LRMP - lymphoid restricted membrane protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204126.1 | 541 | UTR 5 | NP_001191055.1 | |||
NM_001204127.1 | 541 | UTR 5 | NP_001191056.1 | |||
NM_006152.3 | 541 | UTR 5 | NP_006143.2 | |||
XM_005253370.3 | 541 | UTR 5 | XP_005253427.1 | |||
XM_005253372.2 | 541 | UTR 5 | XP_005253429.1 | |||
XM_005253374.3 | 541 | UTR 5 | XP_005253431.1 | |||
XM_006719076.2 | 541 | Intron | XP_006719139.1 | |||
XM_011520667.1 | 541 | UTR 5 | XP_011518969.1 | |||
XM_011520668.2 | 541 | Intron | XP_011518970.1 | |||
XM_011520669.2 | 541 | Intron | XP_011518971.1 | |||
XM_011520670.1 | 541 | Intron | XP_011518972.1 | |||
XM_017019300.1 | 541 | UTR 5 | XP_016874789.1 | |||
XM_017019301.1 | 541 | UTR 5 | XP_016874790.1 | |||
XM_017019302.1 | 541 | UTR 5 | XP_016874791.1 |
Set Membership: |
HapMap |