Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
49 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606401 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPN9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CAPN9 - calpain 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319676.1 | 39 | Intron | NP_001306605.1 | |||
NM_006615.2 | 39 | Intron | NP_006606.1 | |||
NM_016452.2 | 39 | Intron | NP_057536.1 | |||
XM_011544017.1 | 39 | Intron | XP_011542319.1 | |||
XM_011544018.1 | 39 | Intron | XP_011542320.1 | |||
XM_011544019.2 | 39 | UTR 5 | XP_011542321.1 | |||
XM_011544020.1 | 39 | Intron | XP_011542322.1 | |||
XM_017000098.1 | 39 | Intron | XP_016855587.1 | |||
XM_017000099.1 | 39 | Intron | XP_016855588.1 |
Set Membership: |
HapMap |