Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606248 MIM: 182160 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
QPRT PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
QPRT - quinolinate phosphoribosyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318249.1 | Intron | NP_001305178.1 | ||||
NM_001318250.1 | Intron | NP_001305179.1 | ||||
NM_014298.4 | Intron | NP_055113.2 | ||||
XM_005255223.3 | Intron | XP_005255280.2 | ||||
XM_017023101.1 | Intron | XP_016878590.1 |
SPN - sialophorin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |