Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
52 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612604 MIM: 612605 MIM: 612606 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LCE1B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LCE1B - late cornified envelope 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LCE1C - late cornified envelope 1C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276331.1 | 342 | UTR 3 | NP_001263260.1 | |||
NM_178351.3 | 342 | UTR 3 | NP_848128.1 |
LCE1D - late cornified envelope 1D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |