Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
59 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609973 MIM: 609712 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HCN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
HCN3 - hyperpolarization activated cyclic nucleotide gated potassium channel 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020897.2 | 1968 | Intron | NP_065948.1 | |||
XM_011509816.2 | 1968 | Intron | XP_011508118.1 | |||
XM_011509817.2 | 1968 | Intron | XP_011508119.1 | |||
XM_011509818.2 | 1968 | Intron | XP_011508120.1 | |||
XM_017001918.1 | 1968 | Intron | XP_016857407.1 |
PKLR - pyruvate kinase, liver and RBC | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000298.5 | 1968 | UTR 3 | NP_000289.1 | |||
NM_181871.3 | 1968 | UTR 3 | NP_870986.1 | |||
XM_006711386.3 | 1968 | UTR 3 | XP_006711449.1 | |||
XM_011509640.2 | 1968 | UTR 3 | XP_011507942.1 | |||
XM_017001493.1 | 1968 | Intron | XP_016856982.1 |
Set Membership: |
HapMap Validated |