Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
50 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609973 MIM: 609712 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HCN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HCN3 - hyperpolarization activated cyclic nucleotide gated potassium channel 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020897.2 | 2770 | UTR 3 | NP_065948.1 | |||
XM_011509816.2 | 2770 | UTR 3 | XP_011508118.1 | |||
XM_011509817.2 | 2770 | UTR 3 | XP_011508119.1 | |||
XM_011509818.2 | 2770 | UTR 3 | XP_011508120.1 | |||
XM_017001918.1 | 2770 | UTR 3 | XP_016857407.1 |
PKLR - pyruvate kinase, liver and RBC | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000298.5 | 2770 | UTR 3 | NP_000289.1 | |||
NM_181871.3 | 2770 | UTR 3 | NP_870986.1 | |||
XM_006711386.3 | 2770 | Intron | XP_006711449.1 | |||
XM_011509640.2 | 2770 | Intron | XP_011507942.1 | |||
XM_017001493.1 | 2770 | Intron | XP_016856982.1 |
Set Membership: |
JSNP Validated |