Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
51 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600759 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSEN2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PSEN2 - presenilin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000447.2 | 474 | UTR 5 | NP_000438.2 | |||
NM_012486.2 | 474 | UTR 5 | NP_036618.2 | |||
XM_005273199.3 | 474 | Intron | XP_005273256.1 | |||
XM_017001835.1 | 474 | UTR 5 | XP_016857324.1 | |||
XM_017001836.1 | 474 | Intron | XP_016857325.1 |
Set Membership: |
HapMap JSNP |