Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
37 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603412 MIM: 606980 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDC42BPA PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CDC42BPA - CDC42 binding protein kinase alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003607.3 | 10645 | UTR 3 | NP_003598.2 | |||
NM_014826.4 | 10645 | UTR 3 | NP_055641.3 | |||
XM_005273317.3 | 10645 | Intron | XP_005273374.1 | |||
XM_005273318.3 | 10645 | Intron | XP_005273375.1 | |||
XM_005273320.3 | 10645 | Intron | XP_005273377.1 | |||
XM_005273321.3 | 10645 | UTR 3 | XP_005273378.1 | |||
XM_005273322.3 | 10645 | UTR 3 | XP_005273379.1 | |||
XM_005273324.3 | 10645 | UTR 3 | XP_005273381.1 | |||
XM_006711834.3 | 10645 | Intron | XP_006711897.1 | |||
XM_006711835.3 | 10645 | Intron | XP_006711898.1 | |||
XM_011544306.2 | 10645 | Intron | XP_011542608.1 | |||
XM_011544307.2 | 10645 | Intron | XP_011542609.1 | |||
XM_011544308.2 | 10645 | UTR 3 | XP_011542610.1 | |||
XM_011544309.2 | 10645 | Intron | XP_011542611.1 | |||
XM_017002578.1 | 10645 | UTR 3 | XP_016858067.1 | |||
XM_017002579.1 | 10645 | Intron | XP_016858068.1 | |||
XM_017002580.1 | 10645 | Intron | XP_016858069.1 | |||
XM_017002581.1 | 10645 | Intron | XP_016858070.1 | |||
XM_017002582.1 | 10645 | Intron | XP_016858071.1 |
COQ8A - coenzyme Q8A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |