Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613481 MIM: 604550 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC62 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CCDC62 - coiled-coil domain containing 62 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_201435.4 | 416 | Intron | NP_958843.2 | |||
XM_006719643.1 | 416 | Intron | XP_006719706.1 | |||
XM_006719644.1 | 416 | Intron | XP_006719707.1 | |||
XM_011538856.1 | 416 | Intron | XP_011537158.1 | |||
XM_011538857.1 | 416 | Intron | XP_011537159.1 | |||
XM_011538859.1 | 416 | Intron | XP_011537161.1 | |||
XM_017020033.1 | 416 | UTR 5 | XP_016875522.1 |
DENR - density regulated re-initiation and release factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |