Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609457 MIM: 151570 | ||||||||||||||||||||
Literature Links: |
HAL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HAL - histidine ammonia-lyase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LTA4H - leukotriene A4 hydrolase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000895.2 | 1848 | Missense Mutation | ACT,AGT | T,S 600 | NP_000886.1 | |
NM_001256643.1 | 1848 | Missense Mutation | ACT,AGT | T,S 576 | NP_001243572.1 | |
NM_001256644.1 | 1848 | UTR 3 | NP_001243573.1 | |||
XM_005268871.1 | 1848 | UTR 3 | XP_005268928.1 | |||
XM_011538348.1 | 1848 | Intron | XP_011536650.1 | |||
XM_011538349.2 | 1848 | Intron | XP_011536651.1 |