Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 140100 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HP - haptoglobin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TXNL4B - thioredoxin like 4B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142317.1 | Intron | NP_001135789.1 | ||||
NM_001142318.1 | Intron | NP_001135790.1 | ||||
NM_001324354.1 | Intron | NP_001311283.1 | ||||
NM_001324355.1 | Intron | NP_001311284.1 | ||||
NM_017853.2 | Intron | NP_060323.1 | ||||
XM_017023377.1 | Intron | XP_016878866.1 |
Set Membership: |
HapMap |