Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609604 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP1LC3B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MAP1LC3B - microtubule associated protein 1 light chain 3 beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZCCHC14 - zinc finger CCHC-type containing 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015144.2 | 3029 | UTR 3 | NP_055959.1 | |||
XM_005255858.3 | 3029 | Intron | XP_005255915.2 | |||
XM_011522964.2 | 3029 | Intron | XP_011521266.1 | |||
XM_017023082.1 | 3029 | Intron | XP_016878571.1 |