Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604728 | ||||||||||||||||||||
Literature Links: |
USP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
USP3 - ubiquitin specific peptidase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256702.1 | Intron | NP_001243631.1 | ||||
NM_006537.3 | Intron | NP_006528.2 | ||||
XM_017022763.1 | Intron | XP_016878252.1 | ||||
XM_017022764.1 | Intron | XP_016878253.1 | ||||
XM_017022765.1 | Intron | XP_016878254.1 | ||||
XM_017022766.1 | Intron | XP_016878255.1 | ||||
XM_017022767.1 | Intron | XP_016878256.1 | ||||
XM_017022768.1 | Intron | XP_016878257.1 |