Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C6orf163 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C6orf163 - chromosome 6 open reading frame 163 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010868.2 | Intron | NP_001010868.2 | ||||
XM_005248673.3 | Intron | XP_005248730.1 | ||||
XM_006715370.3 | Intron | XP_006715433.1 | ||||
XM_006715372.2 | Intron | XP_006715435.1 | ||||
XM_011535539.1 | Intron | XP_011533841.1 | ||||
XM_017010367.1 | Intron | XP_016865856.1 |
SMIM8 - small integral membrane protein 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |