Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608714 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNTG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SNTG1 - syntrophin gamma 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287813.2 | 444 | Intron | NP_001274742.1 | |||
NM_001287814.2 | 444 | UTR 5 | NP_001274743.1 | |||
NM_001321773.1 | 444 | UTR 5 | NP_001308702.1 | |||
NM_001321775.1 | 444 | Intron | NP_001308704.1 | |||
NM_001321776.1 | 444 | UTR 5 | NP_001308705.1 | |||
NM_001321777.1 | 444 | UTR 5 | NP_001308706.1 | |||
NM_001321778.1 | 444 | Intron | NP_001308707.1 | |||
NM_018967.4 | 444 | UTR 5 | NP_061840.1 | |||
XM_011517548.2 | 444 | UTR 5 | XP_011515850.1 | |||
XM_017013579.1 | 444 | UTR 5 | XP_016869068.1 | |||
XM_017013580.1 | 444 | Intron | XP_016869069.1 | |||
XM_017013581.1 | 444 | UTR 5 | XP_016869070.1 | |||
XM_017013582.1 | 444 | UTR 5 | XP_016869071.1 |