Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCCTCAGGACGTGGGCACGTTGT[C/G]GTCCAGAGAGCAAGAGCGTCGCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614148 MIM: 602241 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1QTNF9B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
C1QTNF9B - C1q and tumor necrosis factor related protein 9B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C1QTNF9B-AS1 - C1QTNF9B antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014442.2 | Intron | NP_001014442.2 | ||||
NM_001135816.1 | Intron | NP_001129288.1 |
MIPEP - mitochondrial intermediate peptidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005932.3 | Intron | NP_005923.2 | ||||
XM_011535097.2 | Intron | XP_011533399.1 | ||||
XM_011535098.2 | Intron | XP_011533400.1 |
Set Membership: |
HapMap Validated |