Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600509 MIM: 600937 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCC8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ABCC8 - ATP binding cassette subfamily C member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000352.4 | Intron | NP_000343.2 | ||||
NM_001287174.1 | Intron | NP_001274103.1 | ||||
XM_011520331.2 | Intron | XP_011518633.1 | ||||
XM_017018197.1 | Intron | XP_016873686.1 | ||||
XM_017018198.1 | Intron | XP_016873687.1 | ||||
XM_017018199.1 | Intron | XP_016873688.1 | ||||
XM_017018200.1 | Intron | XP_016873689.1 | ||||
XM_017018201.1 | Intron | XP_016873690.1 | ||||
XM_017018202.1 | Intron | XP_016873691.1 | ||||
XM_017018203.1 | Intron | XP_016873692.1 | ||||
XM_017018204.1 | Intron | XP_016873693.1 |
KCNJ11 - potassium voltage-gated channel subfamily J member 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |