Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGGACCCCCGATCCACTAACCT[C/T]TTCTCCCCCATTGCGCTGACAGTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602519 | ||||||||||||||||||||
Literature Links: |
LOC101929989 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101929989 - lipopolysaccharide-induced tumor necrosis factor-alpha factor-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP7 - ubiquitin specific peptidase 7 (herpes virus-associated) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286457.1 | 3785 | UTR 3 | NP_001273386.1 | |||
NM_001286458.1 | 3785 | UTR 3 | NP_001273387.1 | |||
NM_001321858.1 | 3785 | UTR 3 | NP_001308787.1 | |||
NM_003470.2 | 3785 | UTR 3 | NP_003461.2 | |||
XM_017023652.1 | 3785 | UTR 3 | XP_016879141.1 | |||
XM_017023653.1 | 3785 | UTR 3 | XP_016879142.1 |