Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTAAGGAGGCCCACACCCTGACTG[A/C]GCCTCAGAGCCACTGCCCTCACAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602519 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929989 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC101929989 - lipopolysaccharide-induced tumor necrosis factor-alpha factor-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP7 - ubiquitin specific peptidase 7 (herpes virus-associated) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286457.1 | 4101 | Intron | NP_001273386.1 | |||
NM_001286458.1 | 4101 | Intron | NP_001273387.1 | |||
NM_001321858.1 | 4101 | Intron | NP_001308787.1 | |||
NM_003470.2 | 4101 | Intron | NP_003461.2 | |||
XM_017023652.1 | 4101 | UTR 3 | XP_016879141.1 | |||
XM_017023653.1 | 4101 | UTR 3 | XP_016879142.1 |
Set Membership: |
HapMap |