Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGGAGGCGGTACCTTCAGGGGAC[A/G]CTCAGGAGTCACTGCACATTAAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603436 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF205 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ZNF205 - zinc finger protein 205 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042428.1 | 297 | Missense Mutation | ACT,GCT | T,A 43 | NP_001035893.1 | |
NM_001278158.1 | 297 | Missense Mutation | ACT,GCT | T,A 43 | NP_001265087.1 | |
NM_003456.2 | 297 | Missense Mutation | ACT,GCT | T,A 43 | NP_003447.2 | |
XM_005255558.2 | 297 | Missense Mutation | ACT,GCT | T,A 43 | XP_005255615.1 |
ZNF205-AS1 - ZNF205 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |