Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCTGCTGCCCGCCGAGAAGTTGC[C/G]GAGGCCAGATTCGATCTTGTTGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612426 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105371083 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC105371083 - uncharacterized LOC105371083 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RMI2 - RecQ mediated genome instability 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152308.2 | Intron | NP_689521.1 |