Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615020 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELP6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ELP6 - elongator acetyltransferase complex subunit 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031703.2 | 1386 | UTR 3 | NP_001026873.2 | |||
XM_005265241.3 | 1386 | UTR 3 | XP_005265298.1 | |||
XM_011533851.2 | 1386 | Intron | XP_011532153.1 | |||
XM_011533852.2 | 1386 | Intron | XP_011532154.1 | |||
XM_011533853.2 | 1386 | Intron | XP_011532155.1 | |||
XM_011533854.2 | 1386 | Intron | XP_011532156.1 | |||
XM_011533856.2 | 1386 | Intron | XP_011532158.1 | |||
XM_011533857.2 | 1386 | Intron | XP_011532159.1 | |||
XM_017006669.1 | 1386 | UTR 3 | XP_016862158.1 | |||
XM_017006670.1 | 1386 | UTR 3 | XP_016862159.1 |