Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF445 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
ZNF445 - zinc finger protein 445 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181489.5 | 7399 | UTR 3 | NP_852466.1 | |||
XM_005265102.2 | 7399 | UTR 3 | XP_005265159.1 | |||
XM_005265103.3 | 7399 | UTR 3 | XP_005265160.1 | |||
XM_011533674.2 | 7399 | UTR 3 | XP_011531976.1 | |||
XM_011533675.2 | 7399 | UTR 3 | XP_011531977.1 | |||
XM_017006345.1 | 7399 | UTR 3 | XP_016861834.1 | |||
XM_017006346.1 | 7399 | UTR 3 | XP_016861835.1 |
Set Membership: |
HapMap Validated |