Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614169 MIM: 612778 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NBEAL2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NBEAL2 - neurobeachin like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NRADDP - neurotrophin receptor associated death domain, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SETD2 - SET domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014159.6 | Intron | NP_054878.5 | ||||
XM_011533631.2 | Intron | XP_011531933.1 | ||||
XM_011533632.2 | Intron | XP_011531934.1 | ||||
XM_011533633.2 | Intron | XP_011531935.1 | ||||
XM_011533634.1 | Intron | XP_011531936.1 | ||||
XM_017006270.1 | Intron | XP_016861759.1 |
Set Membership: |
HapMap |