Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603865 MIM: 602896 | ||||||||||||||||||||
Literature Links: |
GFPT2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GFPT2 - glutamine-fructose-6-phosphate transaminase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005110.3 | 2798 | UTR 3 | NP_005101.1 | |||
XM_006714942.2 | 2798 | UTR 3 | XP_006715005.1 | |||
XM_017010100.1 | 2798 | UTR 3 | XP_016865589.1 |
MAPK9 - mitogen-activated protein kinase 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |