Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300137 | ||||||||||||||||||||
Literature Links: |
IGSF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IGSF1 - immunoglobulin superfamily member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170961.1 | 3574 | Silent Mutation | GTC,GTG | V,V 1102 | NP_001164432.1 | |
NM_001170962.1 | 3574 | Silent Mutation | GTC,GTG | V,V 1088 | NP_001164433.1 | |
NM_001170963.1 | 3574 | Intron | NP_001164434.1 | |||
NM_001555.4 | 3574 | Silent Mutation | GTC,GTG | V,V 1097 | NP_001546.2 | |
NM_205833.3 | 3574 | Intron | NP_991402.1 | |||
XM_011531330.1 | 3574 | Silent Mutation | GTC,GTG | V,V 1102 | XP_011529632.1 | |
XM_011531333.1 | 3574 | Intron | XP_011529635.1 | |||
XM_011531334.1 | 3574 | Intron | XP_011529636.1 |