Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTGAGATGGGTTTCCCATGGCAA[C/G]CTGAATTCCTGCCCCACTCGAGATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612873 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLJ16779 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FLJ16779 - uncharacterized LOC100192386 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC107985425 - uncharacterized LOC107985425 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NKAIN4 - Na+/K+ transporting ATPase interacting 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152864.3 | Intron | NP_690603.3 | ||||
XM_005260192.1 | Intron | XP_005260249.1 | ||||
XM_005260194.3 | Intron | XP_005260251.1 | ||||
XM_011528527.1 | Intron | XP_011526829.1 | ||||
XM_011528528.2 | Intron | XP_011526830.1 | ||||
XM_011528529.2 | Intron | XP_011526831.1 | ||||
XM_017027636.1 | Intron | XP_016883125.1 | ||||
XM_017027637.1 | Intron | XP_016883126.1 | ||||
XM_017027638.1 | Intron | XP_016883127.1 | ||||
XM_017027639.1 | Intron | XP_016883128.1 |