Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGTCCATGCTGCACAGGTGCCG[C/T]GGGCTCCACAGAGCCCTGCCTGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613004 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HTT PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HTT - huntingtin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MSANTD1 - Myb/SANT DNA binding domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042690.1 | 4374 | Intron | NP_001036155.1 | |||
XM_011513465.2 | 4374 | Intron | XP_011511767.1 | |||
XM_011513466.2 | 4374 | Intron | XP_011511768.1 | |||
XM_011513467.2 | 4374 | UTR 5 | XP_011511769.1 | |||
XM_017008169.1 | 4374 | Intron | XP_016863658.1 |